β-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei
Synonym(s):
(R)-3-Hydroxybutanoate:NAD+ oxidoreductase, 3-HBDH
CAS Number:
Products
Brand
Biological Source
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing