Skip to Content
Merck
Benthiavalicarb isopropyl PESTANAL®, analytical standard

Benthiavalicarb isopropyl

Benthiavalicarb isopropyl PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C18H24FN3O3S
CAS Number:
Molecular Weight:
381.46

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing