![Benserazide for peak identification A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/421/251/e4f3ff96-34df-4c27-8dfc-eefb0d254cb3/640/e4f3ff96-34df-4c27-8dfc-eefb0d254cb3.png)
Benserazide for peak identification A
![Benserazide for peak identification A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/421/251/e4f3ff96-34df-4c27-8dfc-eefb0d254cb3/640/e4f3ff96-34df-4c27-8dfc-eefb0d254cb3.png)
Synonym(s):
Benserazide hydrochloride, DL-Serine 2-(2,3,4-trihydroxybenzyl)hydrazide hydrochloride
Empirical Formula (Hill Notation):
C10H15N3O5 · HCl
CAS Number:
Molecular Weight:
293.70
Products
Formula Weight
Gene Alias
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing