Azithromycin N-oxide
Empirical Formula (Hill Notation):
C38H72N2O13
CAS Number:
Molecular Weight:
764.98
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2