Skip to Content
Merck
Anti-ZC3H13 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-ZC3H13 antibody produced in rabbit

Anti-ZC3H13 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-DKFZp434D1812, Anti-KIAA0853, Anti-zinc finger CCCH-type containing 13, KIAA0853, ZC3H13, Zinc finger CCCH domain-containing protein 13

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique