
Anti-SLC19A2 antibody produced in rabbit

Synonym(s):
Anti-Solute carrier family 19 member 2 antibody produced in rabbit, Anti-TC1 antibody produced in rabbit, Anti-ThTr-1 antibody produced in rabbit, Anti-ThTr1 antibody produced in rabbit, Anti-Thiamine carrier 1 antibody produced in rabbit, Anti-Thiamine transporter 1 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1