Anti-PABPN1 antibody produced in rabbit
Synonym(s):
Anti-OPMD, Anti-PAB2, Anti-PABP2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2