
Anti-KNG1 antibody produced in rabbit

Synonym(s):
Anti-BDK, Anti-KNG, Anti-Kininogen 1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing