Anemosapogenin
Synonym(s):
23-Hydroxybetulinic acid, (3β,4α)-3,23-Dihydroxylup-20(29)-en-28-oic acid, 3β,23-Dihydroxylup-20(29)-en-28-oic acid, 3aH-Cyclopenta[a]chrysene, lup-20(29)-en-28-oic acid deriv., Anemosapogenin
Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing