![Amoxicillin trihydrate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/269/072/6a976d6c-471f-4855-b9d2-2ce7564be542/640/6a976d6c-471f-4855-b9d2-2ce7564be542.png)
Amoxicillin trihydrate
![Amoxicillin trihydrate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/269/072/6a976d6c-471f-4855-b9d2-2ce7564be542/640/6a976d6c-471f-4855-b9d2-2ce7564be542.png)
Empirical Formula (Hill Notation):
C16H19N3O5S · 3H2O
CAS Number:
Molecular Weight:
419.45
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing