Skip to Content
Merck
Amiprofos methyl PESTANAL®, analytical standard

Amiprofos methyl

Amiprofos methyl PESTANAL®, analytical standard
Synonym(s):
O-Methyl O-(2-nitro-p-tolyl) N-isopropylphosphoramidothionate
Empirical Formula (Hill Notation):
C11H17N2O4PS
CAS Number:
Molecular Weight:
304.30
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Special Grade