![Aminobutanol United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/391/125/3e244c58-bc03-4cf9-82fd-4692df877f21/640/3e244c58-bc03-4cf9-82fd-4692df877f21.png)
Aminobutanol
![Aminobutanol United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/391/125/3e244c58-bc03-4cf9-82fd-4692df877f21/640/3e244c58-bc03-4cf9-82fd-4692df877f21.png)
Synonym(s):
2-Amino-1-butanol
Linear Formula:
C2H5CH(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing