![Amifostine Disulfide Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/333/909/4a0a2184-4174-4635-bb2d-900751acec7e/640/4a0a2184-4174-4635-bb2d-900751acec7e.jpg)
Amifostine disulfide
![Amifostine Disulfide Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/333/909/4a0a2184-4174-4635-bb2d-900751acec7e/640/4a0a2184-4174-4635-bb2d-900751acec7e.jpg)
Synonym(s):
N,N′-(Dithiodi-2,1-ethanediyl)bis-1,3-propanediamine tetrahydrochloride, WR-33278 tetrahydrochloride
Empirical Formula (Hill Notation):
C10H26N4S2 · 4HCl
CAS Number:
Molecular Weight:
412.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2