![α-Ketoglutaric acid sodium salt BioUltra](/deepweb/assets/sigmaaldrich/product/structures/336/307/93e923eb-3316-41be-a287-d57b878c4963/640/93e923eb-3316-41be-a287-d57b878c4963.png)
α-Ketoglutaric acid sodium salt
![α-Ketoglutaric acid sodium salt BioUltra](/deepweb/assets/sigmaaldrich/product/structures/336/307/93e923eb-3316-41be-a287-d57b878c4963/640/93e923eb-3316-41be-a287-d57b878c4963.png)
Synonym(s):
2-Oxoglutaric acid monosodium salt, 2-Oxopentanedioic acid monosodium salt, Sodium 2-oxoglutarate monobasic
Linear Formula:
HOOCCH2CH2COCOONa
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing