![Aciclovir British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/817/179/cdc344db-5da0-49c9-a29b-b025ce3c722e/640/cdc344db-5da0-49c9-a29b-b025ce3c722e.png)
Aciclovir
![Aciclovir British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/817/179/cdc344db-5da0-49c9-a29b-b025ce3c722e/640/cdc344db-5da0-49c9-a29b-b025ce3c722e.png)
Synonym(s):
Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir
Empirical Formula (Hill Notation):
C8H11N5O3
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing