Skip to Content
Merck
6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester suitable for fluorescence, ≥75% (HPLC)

6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester

6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester suitable for fluorescence, ≥75% (HPLC)
Synonym(s):
Fluorescein-5(6)-carboxamidocaproic acid N-succinimidyl ester
Empirical Formula (Hill Notation):
C31H26N2O10
CAS Number:
Molecular Weight:
586.55

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment