Skip to Content
Merck
vibrant-m

(5RS)-3-(2-Hydroxyphenyl)-5-phenylcyclohex-2-enone

vibrant-m
Synonym(s):
3-(2-Hydroxyphenyl)-5-phenyl-2-cyclohexen-1-one, Alice’s ketone
Empirical Formula (Hill Notation):
C18H16O2
CAS Number:
Molecular Weight:
264.32

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1