![4-Methylumbelliferyl β-D-lactopyranoside β-galactosidase substrate](/deepweb/assets/sigmaaldrich/product/structures/365/295/9ba52d7c-9b4d-460b-a86d-7e9e0b1e64bc/640/9ba52d7c-9b4d-460b-a86d-7e9e0b1e64bc.png)
4-Methylumbelliferyl β-D-lactopyranoside
![4-Methylumbelliferyl β-D-lactopyranoside β-galactosidase substrate](/deepweb/assets/sigmaaldrich/product/structures/365/295/9ba52d7c-9b4d-460b-a86d-7e9e0b1e64bc/640/9ba52d7c-9b4d-460b-a86d-7e9e0b1e64bc.png)
Synonym(s):
4-Methylumbelliferyl β-D-lactoside
Empirical Formula (Hill Notation):
C22H28O13
CAS Number:
Molecular Weight:
500.45
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing