Skip to Content
Merck
vibrant-m

3-Piperidylpropiophenone hydrochloride

vibrant-m
Synonym(s):
1-Phenyl-3-(piperidin-1-yl)propan-1-one hydrochloride, 3-Piperidinopropiophenone hydrochloride
Empirical Formula (Hill Notation):
C14H19NO · HCl
CAS Number:
Molecular Weight:
253.77

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1