
3-Methylflavone-8-carboxylic acid

Synonym(s):
3-Methylflavone-8-carboxylic acid, 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid
Empirical Formula (Hill Notation):
C17H12O4
CAS Number:
Molecular Weight:
280.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing