Skip to Content
Merck
2-Iodopropane contains copper as stabilizer, 99%

2-Iodopropane

2-Iodopropane contains copper as stabilizer, 99%
Synonym(s):
Isopropyl iodide, 2-Iodopropane
Linear Formula:
(CH3)2CHI
CAS Number:
Molecular Weight:
169.99

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing