![2-Hydroxycyclohexanone dimer 97%](/deepweb/assets/sigmaaldrich/product/structures/204/742/d3274011-d8ff-4a58-9efb-98236ef948ef/640/d3274011-d8ff-4a58-9efb-98236ef948ef.png)
2-Hydroxycyclohexanone dimer
![2-Hydroxycyclohexanone dimer 97%](/deepweb/assets/sigmaaldrich/product/structures/204/742/d3274011-d8ff-4a58-9efb-98236ef948ef/640/d3274011-d8ff-4a58-9efb-98236ef948ef.png)
Synonym(s):
Adipoin, NSC 117392
Linear Formula:
[HOC6H9(=O)]2
CAS Number:
Molecular Weight:
228.28
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing