Skip to Content
Merck
2-Furyl methyl ketone natural (US), ≥97%, FG

2-Furyl methyl ketone

2-Furyl methyl ketone natural (US), ≥97%, FG
Synonym(s):
2-Acetylfuran
Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4