2-Chloro-p-phenylenediamine monosulfate
Synonym(s):
2-Chloro-1,4-diaminobenzene monosulfate
Empirical Formula (Hill Notation):
C6H7ClN2 · H2SO4
CAS Number:
Molecular Weight:
240.66
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing