Skip to Content
Merck
2-Chloroadenine 96%

2-Chloroadenine

2-Chloroadenine 96%
Synonym(s):
2-Chloro-6-aminopurine
Empirical Formula (Hill Notation):
C5H4ClN5
CAS Number:
Molecular Weight:
169.57

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing