![2-Aminoacridone BioReagent, suitable for fluorescence, ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/297/355/1abc5b7c-a2a5-48d0-aac6-bafb30814cf2/640/1abc5b7c-a2a5-48d0-aac6-bafb30814cf2.png)
2-Aminoacridone
![2-Aminoacridone BioReagent, suitable for fluorescence, ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/297/355/1abc5b7c-a2a5-48d0-aac6-bafb30814cf2/640/1abc5b7c-a2a5-48d0-aac6-bafb30814cf2.png)
Synonym(s):
2-Amino-9(10H)-acridinone, 2-amino-10H-acridin-9-one, AMAC
Empirical Formula (Hill Notation):
C13H10N2O
CAS Number:
Molecular Weight:
210.23
Products
Product Category
Brand
Formula Weight
Product Line
Purity
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing