Skip to Content
Merck
(1R)-(+)-α-Pinene 97%, FG

(1R)-(+)-α-Pinene

(1R)-(+)-α-Pinene 97%, FG
Synonym(s):
(+)-α-Pinene, (1R,5R)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1R,5R)-2-Pinene, (1R)-(+)-α-Pinene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing