Skip to Content
Merck
1-Penten-3-one contains 0.1% BHT as stabilizer, 97%

1-Penten-3-one

1-Penten-3-one contains 0.1% BHT as stabilizer, 97%
Synonym(s):
Ethyl vinyl ketone
Linear Formula:
C2H5COCH=CH2
CAS Number:
Molecular Weight:
84.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing