Skip to Content
Merck
[1,3-Bis(diphenylphosphino)propane]dichloronickel(II)

[1,3-Bis(diphenylphosphino)propane]dichloronickel(II)

[1,3-Bis(diphenylphosphino)propane]dichloronickel(II)
Synonym(s):
(1,3-dppp)NiCl2, 1,3-Bis(diphenylphosphino)propane nickel(II) chloride, Ni(dppp)Cl2
Linear Formula:
[(C6H5)2P(CH2)3P(C6H5)2]NiCl2
CAS Number:
Molecular Weight:
542.04

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing