1,2,4-Triazole
Synonym(s):
1,2,4-Triazole
Empirical Formula (Hill Notation):
C2H3N3
CAS Number:
Molecular Weight:
69.07
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3