Product Name
BLUeye Prestained Protein Ladder room temperature stable molecular weight markers
Product Number
94964
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Report Results
APPEARANCE (FORM)Liquid
SDS GEL ELECTROPHORESIS12 prestained proteins (native molecular weight of) 5.6 kda 10.5 kDa 14.5 kDa 21.1 kDa 30.1 kDa 41.9 kDa 54 kDa 70.3 kDa 92 kDa132 kDa 175 kDa 235 kDa