Product Name
Sodium methanesulfinate technical grade, 85%
Product Number
433063
Product Brand
aldrich
CAS Number
20277-69-4
Molecular Weight
102.09
Test
Specification
Appearance (Form)Conforms to Requirements
 Powder and/or Chunks
Infrared spectrumConforms to Structure
Purity (Titration by HClO4)≥82.5 %