Skip to Content
Merck
All Photos(3)

Key Documents

V900388

Sigma-Aldrich

Sodium deoxycholate

Vetec, reagent grade, ≥97%

Synonym(s):

3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NaO4
CAS Number:
Molecular Weight:
414.55
Beilstein:
3581950
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥97%

form

powder

mol wt

micellar avg mol wt 1200-5000

aggregation number

3-12

technique(s)

protein quantification: suitable

CMC

2-6 mM (20-25°C)

HLB

16

functional group

carboxylic acid

storage temp.

room temp

SMILES string

[Na+].[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC([O-])=O)[C@@]4(C)[C@@H](O)C[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C24H40O4.Na/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;/m1./s1

InChI key

FHHPUSMSKHSNKW-SMOYURAASA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L L Liu et al.
Oncogene, 33(24), 3140-3150 (2013-07-16)
Prostate tumors develop resistance to androgen deprivation therapy (ADT) by multiple mechanisms, one of which is to express constitutively active androgen receptor (AR) splice variants lacking the ligand-binding domain. AR splice variant 7 (AR-V7, also termed AR3) is the most
Shin Yoshimoto et al.
Nature, 499(7456), 97-101 (2013-06-28)
Obesity has become more prevalent in most developed countries over the past few decades, and is increasingly recognized as a major risk factor for several common types of cancer. As the worldwide obesity epidemic has shown no signs of abating
Lotta K Stenman et al.
American journal of physiology. Gastrointestinal and liver physiology, 304(3), G227-G234 (2012-12-04)
Impairment of gut barrier is associated with a fat-rich diet, but mechanisms are unknown. We have earlier shown that dietary fat modifies fecal bile acids in mice, decreasing the proportion of ursodeoxycholic acid (UDCA) vs. deoxycholic acid (DCA). To clarify
Ayako Suzuki et al.
Nucleic acids research, 42(22), 13557-13572 (2014-11-08)
Here we conducted an integrative multi-omics analysis to understand how cancers harbor various types of aberrations at the genomic, epigenomic and transcriptional levels. In order to elucidate biological relevance of the aberrations and their mutual relations, we performed whole-genome sequencing
Barbara Stefanska et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(12), 3118-3132 (2014-04-26)
We utilized whole-genome mapping of promoters that are activated by DNA hypomethylation in hepatocellular carcinoma (HCC) clinical samples to shortlist novel targets for anticancer therapeutics. We provide a proof of principle of this approach by testing six genes short-listed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service