Skip to Content
Merck
All Photos(1)

Documents

Z740521

Brady® BMP51 printer kit

AC/DC input 100 - 240 V AC, EuroPlug (2-pin)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

feature

Kit includes: BMP51 Printer with battery pack and AC adaptor/battery charger

packaging

pkg of 1 kit

manufacturer/tradename

Brady 710894EU

Looking for similar products? Visit Product Comparison Guide

General description

The BMP51 Label Maker is a compact, versatile portable printer with built-in Bluetooth® technology for wireless printing. When used in conjunction with the free Brady Mobile App, you can design and print labels with this printer using your Android phone!

This label maker has a 300 dpi print head and can print professional, durable labels up to 38.10 mm wide. It prints on continuous and die-cut materials at a speed of 25.40 mm per second. With a built-in cutter, easy top-loading cartridges, and 3 different options for battery power, the BMP51 Label Maker is a powerful printer for on-the-job label making.

The BMP51 has a full QWERTY keyboard, for stand alone operation. The BMP51 Label Maker is ideal for electrical, datacom, facility/safety, product and panel manufacturing, and laboratory labelling applications.

Get the job done faster with more confidence.

  • Change supplies in seconds with hassle-free drop-in cartridges
  • Automatic label formatting – no printer calibration!
  • QWERTY style keypad for faster, more accurate typing
  • Battery options for portability (AA batteries, rechargeable Li-Ion battery or AC power)
  • Labels up to 1.5 in. wide
  • Continuous & die-cut label materials
  • Import, store, and print label files from Excel®
  • Brady labels will not fade, smear or fall off – so you only need to label something once
  • Rugged printer design can survive a 4-foot drop onto concrete, harsh environments and extreme temperatures

Legal Information

Bluetooth is a registered trademark of Bluetooth SIG, Inc.
Brady is a registered trademark of Brady Worldwide, Inc.
Excel is a registered trademark of Microsoft Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service