Skip to Content
Merck
All Photos(1)

Key Documents

T116

Supelco

Thebaine

Synonym(s):

Paramorphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO3
CAS Number:
Molecular Weight:
311.37
EC Number:
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

drug control

USDEA Schedule II; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

[H][C@@]12Oc3c(OC)ccc4C[C@H]5N(C)CC[C@]1(C5=CC=C2OC)c34

InChI

1S/C19H21NO3/c1-20-9-8-19-12-5-7-15(22-3)18(19)23-17-14(21-2)6-4-11(16(17)19)10-13(12)20/h4-7,13,18H,8-10H2,1-3H3/t13-,18+,19+/m1/s1

InChI key

FQXXSQDCDRQNQE-VMDGZTHMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaik B Kok et al.
Organic & biomolecular chemistry, 9(4), 1008-1011 (2010-12-28)
Further investigations into the direct synthesis of N-nororipavine from oripavine using iron powder under non-classical Polonovski conditions have been conducted. The stoichiometry, solvents and iron oxidation rates were found to have a dramatic effect on the rate of N-demethylation as
Xucong Lin et al.
Journal of separation science, 30(17), 3011-3017 (2007-10-27)
A method for the separation and determination of five major opium alkaloids (narcotine, papaverine, thebaine, codeine, and morphine) in pericarpium papaveris by pressurized CEC (pCEC) with monolithic column has been developed. Under the optimum condition, linear calibration ranges of narcotine
Gilbert Stork et al.
Journal of the American Chemical Society, 131(32), 11402-11406 (2009-07-25)
Total syntheses of the morphine alkaloids are described that use a direct stereoselective formation of the phenanthrofuran system via an intramolecular 4 + 2 cycloaddition of a diene tethered to the 4-position of a 7-methoxybenzofuran-3-carboxylic acid ester.
Robert J Carroll et al.
The Journal of organic chemistry, 74(2), 747-752 (2008-12-17)
The ethylene glycol ketal of neopinone was prepared in a one-pot procedure by the reaction of thebaine with ethylene glyocol in the presence of p-toluenesulfonic acid. The ketal is also an intermediate in the conversion of thebaine to hydrocodone with
Ali Reza Fakhari et al.
Journal of separation science, 33(14), 2153-2159 (2010-06-17)
This study investigated the use of ultrasound-assisted extraction to improve the extraction efficiency of morphine, codeine and thebaine from the papaver plants. Extraction conditions such as type of solvent, temperature, duration, frequency and power level of ultrasonic were optimized and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service