Skip to Content
Merck
All Photos(1)

Documents

R8507

Sigma-Aldrich

Hpa I from Haemophilus parainfluenzae

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

grade

for molecular biology

form

buffered aqueous glycerol solution

concentration

3,000-10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Specificity

Recognition sequence: 5′-GTT/AAC-3′
Ligation and recutting results: After 2-10-fold Hpa I overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >80% of fragments can be ligated, and >95% recut.
Heat inactivation: Not completely inactivated at 65 °C for 15 minutes.

Application

HpaI is a restriction endonuclease that is used to cleave DNA at the recognition site 5′-GTT/AAC-3′, generating fragments with blunt ends.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SA (B7531).

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 50 mM KCl, 0.5 mM EDTA, 5 mM 2-mercaptoethanol, 0.01% polydocanol, 50% glycerol (v/v) at 4°C

incubation buffer

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Q Gu et al.
The Journal of general physiology, 118(5), 481-494 (2001-11-07)
Noncovalent molecular adapters, such as cyclodextrins, act as binding sites for channel blockers when lodged in the lumen of the alpha-hemolysin (alphaHL) pore, thereby offering a basis for the detection of a variety of organic molecules with alphaHL as a
Tatiana Gianni et al.
Journal of virology, 78(22), 12268-12276 (2004-10-28)
Herpes simplex virus (HSV) enters cells by fusion with target membranes, commonly the plasma membrane. In some cells, including CHO cells expressing the nectin1 or herpesvirus entry mediator receptors, entry occurs through an endocytic route. We report the following results.
Nucleotide sequences at the cleavage sites of two restriction endonucleases from Hemophilus parainfluenzae.
D E Garfin et al.
Biochemical and biophysical research communications, 59(1), 108-116 (1974-07-10)
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by
Saqib H Ansari et al.
Journal of pediatric hematology/oncology, 35(4), e153-e156 (2013-02-08)
β-thalassemia is characterized by impaired β-chain synthesis leading to ineffective erythropoiesis, severe anemia, and a need for blood transfusion. Presence of Xmn I polymorphism (-158 C-T nucleotide change) in γ-globin gene is associated with a higher fetal hemoglobin and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service