Skip to Content
Merck
All Photos(2)

Key Documents

HPA011173

Sigma-Aldrich

Anti-SRPRB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein APMCF1, Anti-SR-beta, Anti-Signal recognition particle receptor subunit beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LCDSGKTLLFVRLLTGLYRDTQTSITDSCAVYRVNNNRGNSLTLIDLPGHESLRLQFLERFKSSARAIVFVVDSAAFQREVKDVAEFLYQVLIDSMGLKNTPSFLIACNKQDIAMAKSAKLIQQQLEKELNTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SRPRB(58477)

General description

SRPRB (signal recognition particle receptor, B subunit) is a member of the small G-protein superfamily, and was first identified in the cDNA library of breast carcinoma MCF-7 cell line. It is also a signal recognition particle receptor β (SRβ), and is made of 271 amino acids. It has a GTPase domain and resides in the cytoplasm. This gene is localized to human chromosome 3q22.2, spans 14.8kb, and contains seven exons and six introns. It has a transmembrane helix, and is expressed ubiquitously.

Immunogen

Signal recognition particle receptor subunit beta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The function of SRPRB (signal recognition particle receptor, B subunit) is not well characterized. Since, it is a G-protein it might be involved in multiple cellular events such as, cell division and differentiation, vesicular transport, nuclear assembly, and modulation of the cytoskeleton. It is a novel gene related to apoptosis, and is over-expressed in colon cancer. It therefore, might play a role in the control of cell cycle. It is also over-expressed in esophagus, lung, breast and liver carcinomas. It might have a role in tumorigenesis. It controls p21 and TIMP3 (TIMP metallopeptidase inhibitor 3) genes, and thus causes cell cycle arrest at the G1 phase in human hepatocellular carcinoma cells (HHCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72051

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaqing Zhang et al.
Journal of experimental & clinical cancer research : CR, 28, 111-111 (2009-08-12)
APMCF1 is a novel human gene first cloned from apoptotic MCF-7 cells. Our previous study found ectogenic APMCF1 could induce G1 arrest in hepatocarcinoma cell line HHCC. In order to search its broad expression profile for further understanding of its
Qinlong Li et al.
Molecular biology reports, 33(4), 257-263 (2006-11-03)
We previously found that there was up-regulation of APMCF1 expression in apoptotic MCF-7 cells. Moreover, bioinformatics analysis has found that APMCF1 molecules had similar size and structure with molecules which belong to small G-protein superfamily. We presume that APMCF1 plays
Wei Yan et al.
World journal of gastroenterology, 9(8), 1719-1724 (2003-08-15)
APMCF1 is a novel human gene whose transcripts are up-regulated in apoptotic MCF-7 cells. In order to learn more about this gene's function in other tumors, we cloned its full length cDNA and prepared its polyclonal antibody to investigate its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service