Recommended Products
product name
HL60 M4, 88120804
biological source
human blood
growth mode
Suspension
karyotype
Modal no. 45
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian promyelocytic leukaemia
Cell Line Description
HL60 induced to mature into neutrophils with 1.75% DMSO after treating with SV40. Fails to express SV40 specific T Ag suggesting that HL60 M4 represents expansion of sub-clones of HL60 with inherent restricted capacity for neutrophil differentiation.
Application
Differentiation studies
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 1.25% Dimethyl Sulphoxide (DMSO).
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml. Do not allow the culture medium to become acidic or the cells will differentiate; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service