Skip to Content
Merck
All Photos(1)

Documents

C9154

Sigma-Aldrich

Choline base solution

~50 % (w/w) in H2O

Synonym(s):

(2-Hydroxyethyl)trimethylammonium hydroxide, Choline solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(OH)CH2CH2OH
CAS Number:
Molecular Weight:
121.18
Beilstein:
1736748
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

concentration

~50 % (w/w) in H2O

storage temp.

2-8°C

SMILES string

[OH-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.H2O/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H2/q+1;/p-1

InChI key

KIZQNNOULOCVDM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Acyl group acceptor

Caution

May darken in storage.

Substrates

Choline acetyltransferase substrate

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Dan Hou et al.
Biotechnology and bioengineering, 112(1), 65-73 (2014-07-30)
In this work, 28 cholinium ionic liquids (ILs), most of which are good solvents for dissolving lignin, were used for rice straw pretreatment to improve subsequent enzymatic hydrolysis. The anion exerted a significant effect on the pretreatment effectiveness of the
Kyung-Eui Park et al.
Bioscience, biotechnology, and biochemistry, 78(6), 981-988 (2014-07-19)
Phospholipase D (PLD) catalyzes the hydrolysis of phosphatidylcholine (PC), the most abundant phospholipids of plasma membrane, resulting in the production of choline and phosphatidic acid (PA). Choline is a precursor of the neurotransmitter acetylcholine, whereas PA functions as an intracellular
Yan-Xia An et al.
Bioresource technology, 192, 165-171 (2015-06-01)
Pretreatment of lignocelluloses is a key step in the biorefinery for production of biofuels and valuable platform chemicals. In this work, various lignocelluloses were pretreated using cholinium ionic liquids (ILs) that are wholly composed of biomaterials, and fractionated into carbohydrate-rich
Bieke Onghena et al.
Dalton transactions (Cambridge, England : 2003), 43(30), 11566-11578 (2014-06-19)
The ionic liquid choline bis(trifluoromethylsulfonyl)imide, [Chol][Tf2N], was used for the extraction of neodymium(III), in combination with choline hexafluoroacetylacetonate, [Chol][hfac], as the extractant. The binary mixture of [Chol][Tf2N] and water shows temperature-dependent phase behavior, with an upper critical solution temperature of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service