Skip to Content
Merck
All Photos(1)

Documents

C2517

Sigma-Aldrich

Chlordiazepoxide hydrochloride

Synonym(s):

7-Chloro-2-(methylamino)-5-phenyl-3H-1,4-benzodiazepine 4-oxide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClN3O · HCl
CAS Number:
Molecular Weight:
336.22
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

Cl[H].CNC1=Nc2ccc(Cl)cc2C(c3ccccc3)=[N+]([O-])C1

InChI

1S/C16H14ClN3O.ClH/c1-18-15-10-20(21)16(11-5-3-2-4-6-11)13-9-12(17)7-8-14(13)19-15;/h2-9H,10H2,1H3,(H,18,19);1H

InChI key

DMLFJMQTNDSRFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The use of chlordiazepoxide during the first trimester might lead to congenital defects.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Psychologists' psychotropic drug reference. (1999)
J K Pereira et al.
Life sciences, 65(10), PL101-PL107 (1999-09-28)
A single exposure to the elevated plus-maze test of anxiety reduces or abolishes the anxiolytic efficacy of benzodiazepines. The present study was designed to examine whether this phenomenon of "one-trial tolerance" resulted from a motivational deficit on trial 2. We
A Agmo et al.
Neuropharmacology, 37(2), 223-232 (1998-07-29)
Previous studies have shown that the opiate antagonist naloxone blocks the anxiolytic-like effects of benzodiazepines in several models of anxiety, including the elevated plus-maze. Although naloxone preferentially binds to the mu opioid receptor, its selectivity is rather low. The opioid
D J Cash et al.
The Journal of pharmacology and experimental therapeutics, 283(2), 704-711 (1997-11-14)
When rats were made tolerant to the benzodiazepine tranquilizer chlordiazepoxide (CDPX) by its steady administration, a particular gamma-aminobutyric acid type A (GABAA) receptor in cerebral cortex was modified. Its rate of desensitization in the absence of CDPX was enhanced (3-fold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service