Skip to Content
Merck
All Photos(2)

Key Documents

Y22001

Sigma-Aldrich

EnPresso® Y Defined

Growth system for expressing protein in yeast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Pricing and availability is not currently available.

sterility

sterile; γ-irradiated

form

tablet

pH

6.8

shipped in

ambient

storage temp.

room temp

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
Z743838Z742760Z743829
manufacturer/tradename

Molecular BioProducts 3502-RT

manufacturer/tradename

Molecular BioProducts 3552-RT

manufacturer/tradename

Molecular BioProducts 3771-RT

manufacturer/tradename

Molecular BioProducts 3102-RT

feature

ART 10, reload tower

feature

ART 200, reload tower

feature

ART 300, reload tower

feature

ART 1000, reload tower

packaging

pkg of 5X960 ea

packaging

pkg of 5X960 ea

packaging

pkg of 5X960 ea

packaging

pkg of 5X560 ea

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

size

10 μL

size

200 μL

size

300 μL

size

1000 μL

Biochem/physiol Actions

EnPresso® Y Defined is a pre-sterilized growth system designed to increase the yield of functional proteins expressed in Pichia pastoris when using AOX promoters. The system provides stress-free conditions, faster growth and eliminates the need for time-consuming media changes required when using conventional methanol-induction. EnPresso® growth systems provide optimal conditions for growth, metabolism and protein expression in microbial cultures. Protein yields are increased by enabling cultures to reach far higher cell densities than those achieved using conventional media.

By controlling growth rate and metabolism, a greater proportion of expressed protein can be correctly folded to improve solubility, minimize the risk of inclusion body formation, and ensure functionality of the final product.

EnPresso® growth systems maintain pH, provide adequate minerals, vitamins and trace elements to support growth, and use proprietary EnBase technology to ensure a constant, slow release of glucose from a polysaccharide substrate.

See all available products from EnPresso B Growth Systems.

Physical form

EnPresso® B is supplied in a kit providing sufficient reagents for 40 separate 50 ml cultures. Included in the kit:
120 tablets in 40 orange bags
1 bottle (20 ml) Reagent A

Legal Information

EnBase is a trademark of BioSilta Oy
EnPresso is a registered trademark of BioSilta Oy

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service