Skip to Content
Merck
All Photos(1)

Documents

07408

Sigma-Aldrich

N-[Nα,Nα-Bis(carboxymethyl)-L-lysine]-16-mercaptohexadecanamide

for complexometry, 97%

Synonym(s):

N2,N2-Bis(carboxymethyl)-N6-(16-mercapto-1-oxohexadecyl)-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H48N2O7S
CAS Number:
Molecular Weight:
532.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

grade

for complexometry

Assay

≥97% (CHN)
97%

SMILES string

OC(=O)CN(CC(O)=O)[C@@H](CCCCNC(=O)CCCCCCCCCCCCCCCS)C(O)=O

InChI

1S/C26H48N2O7S/c29-23(17-12-10-8-6-4-2-1-3-5-7-9-11-15-19-36)27-18-14-13-16-22(26(34)35)28(20-24(30)31)21-25(32)33/h22,36H,1-21H2,(H,27,29)(H,30,31)(H,32,33)(H,34,35)/t22-/m0/s1

InChI key

GIASYAZZSODPPA-QFIPXVFZSA-N

Application

N-[Nα,Nα-Bis(carboxymethyl)-L-lysine]-16-mercaptohexadecanamide is used in protein separation by linking Ni2+ and Au by coordinating with the nickel and providing a strong interaction betwenn Au-S .

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Currently available only in Europe.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jie Bao et al.
ACS nano, 1(4), 293-298 (2007-11-01)
In this article, we report the synthesis of bifunctional Au-Fe(3)O(4) nanoparticles that are formed by chemical bond linkage. Due to the introduction of Au nanoparticles, the resulting bifunctional Au-Fe(3)O(4) nanoparticles can be easily modified with other functional molecules to realize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service