Skip to Content
Merck
All Photos(1)

Key Documents

NIST2235

Bismuth for thermal analysis

NIST® SRM® 2235

Sign Into View Organizational & Contract Pricing

Select a Size

1.5 G
€531.00

€531.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1.5 G
€531.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€531.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

lumps

packaging

pkg of 1.5 g

manufacturer/tradename

NIST®

application(s)

general analytical

General description

This Standard Reference Material (SRM) is intended for use in calibrating differential scanning calorimeters, differential thermal analyzers, and similar instruments. A unit of SRM 2235 consists of 1.5 g of high purity bismuth in the form of shot with diameters of about 1 mm. For more information, please refer to the SDS and COA.

SRM 2235_cert

SRM 2235_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Bismuth (Bi)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service