OP55
Anti-c-Jun/AP-1 (Ab-3) Mouse mAb (76)
liquid, clone 76, Calbiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
biological source
mouse
Quality Level
antibody product type
primary antibodies
clone
76, monoclonal
form
liquid
does not contain
preservative
species reactivity
human, mouse, rat
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
isotype
IgG1
shipped in
wet ice
storage temp.
−20°C
General description
Purified mouse monoclonal antibody. Recognizes the ~32-39 kDa c-Jun/AP-1 protein.
Recognizes the ~39 kDa c-Jun/AP-1 protein in proliferating cells and most tumor tissues.
Immunogen
recombinant jun protein
Application

Immunoblotting (2.5 g/ml)
Paraffin Sections (2.5 g/ml, pressure cooker pre-treatment required)
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Physical form
In 0.05 M sodium phosphate buffer, 50% glycerol.
Reconstitution
Following initial thaw, aliquot and freeze (-20°C).
Analysis Note
Negative Control
Quiescent cells
Quiescent cells
Positive Control
Growing cells (such as HeLa) or breast or colon tissue
Growing cells (such as HeLa) or breast or colon tissue
Other Notes
This is the preferred c-Jun antibody for immunoblotting. Antibody should be titrated for optimal results in individual systems.
Volm, M., et al. 1993. Clin. Exp. Metastasis11, 325.
Angel, P., et al. 1988. Nature332, 166.
Rouscher III, F.J., et al. 1988. Science240, 1010.
Sassone-Corsi, P., et al. 1988. Cell54, 553.
Bohmann, D., et al. 1987. .Science238, 1386.
Vogt, P.K., et al. 1987. Proc. Natl. Acad. Sci. USA84, 3316.
Arndt, K., et al. 1986. Proc. Natl. Acad. Sci. USA83, 8516.
Cavaleri, F., et al. 1985. Virology143, 680.
Angel, P., et al. 1988. Nature332, 166.
Rouscher III, F.J., et al. 1988. Science240, 1010.
Sassone-Corsi, P., et al. 1988. Cell54, 553.
Bohmann, D., et al. 1987. .Science238, 1386.
Vogt, P.K., et al. 1987. Proc. Natl. Acad. Sci. USA84, 3316.
Arndt, K., et al. 1986. Proc. Natl. Acad. Sci. USA83, 8516.
Cavaleri, F., et al. 1985. Virology143, 680.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yong Jung et al.
International journal of oncology, 40(4), 1122-1132 (2011-12-20)
A set of proteins reflecting the prognosis of patients have clinical significance since they could be utilized as predictive biomarkers and/or potential therapeutic
Peter Lapan et al.
PloS one, 4(8), e6822-e6822 (2009-09-24)
Many important biological processes are controlled through cell-cell interactions, including the colonization of metastatic tumor cells and the control of differentiation of stem cells within their niche. Despite the crucial importance of the cellular environment in regulating cellular signaling, in
Héloïse Macaire et al.
The Journal of biological chemistry, 287(25), 21357-21370 (2012-05-04)
Human T lymphotropic virus type 1 (HTLV-1) is the etiologic agent of adult T-cell leukemia/lymphoma (ATLL). ATLL is a severe malignancy with no effective treatment. HTLV-1 regulatory proteins Tax and HTLV-1 basic leucine zipper factor (HBZ) play a major role
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service