Skip to Content
Merck
All Photos(2)

Documents

MXQCAM001

Millipore

Milliflex® Quantum Camera

For use with Milliflex® Quantum system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.79

material

304L stainless steel plate (front)
aluminum housing
epoxy-coated housing
glass window(s)

manufacturer/tradename

Milliflex®

application(s)

pharmaceutical

compatibility

for use with Milliflex® Quantum Systems

shipped in

ambient

General description

The Camera is designed to be used with the Milliflex® Quantum system. Milliflex® Quantum is a fluorescence-based technology for the quantitative and rapid detection of microorganisms in filterable samples. The universal enzymatic fluorescent staining is non-destructive, allowing the identification of the contaminant following a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms. The Milliflex® Quantum camera combined with the Milliflex® Quantum Spot Counter software makes the sample reading more convenient and allows tracking of records for optimized microbiology QC labs.

Application

The Milliflex® Quantum camera connected to a PC makes sample reading more convenient during the Milliflex® filtration test of filtrable samples, including in-process and final product samples (pharmaceuticals and water monitoring).

Other Notes

Input and Output: Powered by the computer used

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service