Skip to Content
Merck
All Photos(2)

Key Documents

AB5054

Sigma-Aldrich

Anti-Calretinin Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

serum

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... CALB2(794)

General description

Calretinin is a 29 kDa calcium binding protein that is expressed in central and peripheral nervous system and in many normal and pathological tissues. Calretinin is also considered an important diagnostic aid in the differential diagnosis of cystic and solid ameloblastic tumors. The rat and human calretinin exhibit 98% sequence homology and 91% homology to many other species. Calretinin is generally expressed with two other calcium binding proteins Calbindin D-29 and parvalbumin in CNS and other peripheral tissues. Calretinin antibodies have been used to detect a number of carcinomas by clinicopathological, immunohistochemical techniques including, epithelial malignant mesotheliomas, Ovarian yolk sac tumor, Adenomatoid tumors of the uterus etc.

Specificity

Reactivity with other species has not been determined
Specific for calretinin. Recognizes both the calcium-bound and calcium-unbound conformations of calretinin by western blots.

Immunogen

Recombinant rat calretinin.

Application

Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience
This Anti-Calretinin Antibody is validated for use in IH, IH(P), WB for the detection of Calretinin.
Western blot:
1:1,000-1:2,000 of a previous lot was used.

Immunohistochemistry:
1:1,000-1:10,000
Antibody works in paraffin embedded tissues.

Optimal working dilutions must be determined by the end user.

Quality

Immunohistochemistry(paraffin) Analysis:
Calretinin (cat. # AB5054) staining on Human Cerebellum. Tissue pretreated with Citrate, pH 6.0 plus. Antibody was diluted to 1:200 (5.0 µg/mL), using IHC-Select Detection with HRP-DAB. Immunoreactivity is seen as a nuclear and cytoplasmic punctate staining on neurons (brown).
Optimal Staining With Citrate Buffer Epitope Retrieval: Human Cerebellum.

Linkage

Replaces: AB149

Physical form

Rabbit polyclonal serum contains no preservative.
Unpurified

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquots from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Rat brain cortex, Mesothelioma tissue, Human cerebellum.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium-binding Protein Calretinin Immunoreactivity in the Dog Superior Colliculus.
Lee, Jea-Young, et al.
Acta Histochemica Et Cytochemica, 39, 125-138 (2006)
Allison Walter et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(9), 3360-3369 (2010-05-06)
This study evaluated the in vivo antitumor activity of grape-derived polyphenols. BALB/c mice were subcutaneously implanted with C26 colon carcinoma cells, and 2 d later they received either solvent or red wine polyphenols (RWPs) (100 mg/kg/d, human equivalent dose approximately
Repeated phencyclidine in monkeys results in loss of parvalbumin-containing axo-axonic projections in the prefrontal cortex.
Bret A Morrow, John D Elsworth, Robert H Roth
Psychopharmacology (Psychopharmacologia) null
Immunohistochemical and histochemical characteristics of the olfactory system of the guppy, Poecilia reticulata (Teleostei, Poecilidae).
Bettini S, Lazzari M, Ciani F, Franceschini V
Anatomical Record (Hoboken, N.J. : 2007) null
Hippocampal and visuospatial learning defects in mice with a deletion of frizzled 9, a gene in the Williams syndrome deletion interval.
Zhao, C; Aviles, C; Abel, RA; Almli, CR; McQuillen, P; Pleasure, SJ
Development null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service