8.45088
Ethylamine (70% solution in water)
Msynth™plus
Synonym(s):
Aminoethane, Monoethylamine
About This Item
Recommended Products
vapor pressure
460 hPa ( 20 °C)
form
solution
autoignition temp.
375 °C
potency
400 mg/kg LD50, oral (Rat)
265 mg/kg LD50, skin (Rabbit)
expl. lim.
3.0-12.8 % (v/v)
pH
>14 (20 °C in H2O)
bp
39 °C/1013 hPa
mp
<-70 °C
transition temp
flash point -26 °C
density
0.81 g/cm3 at 20 °C
storage temp.
2-30°C
Analysis Note
Appearance of substance (visual): clear
Assay (acidimetric): 68.0 - 72.0 %
Density (d 20 °C/ 4 °C): 0.803 - 0.807
The analysis of residual solvents is not possible due to the physicochemical properties of the material.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(C)
-26 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service