Skip to Content
Merck
All Photos(1)

Key Documents

5.05276

Sigma-Aldrich

HSP70 Activator II, YM-08

Synonym(s):

HSP70 Activator II, YM-08, (2Z,5E)-3-ethyl-5-(3-methylbenzo[d]thiazol-2(3H)-ylidene)-2-(pyridin-2-ylmethylene)thiazolidin-4-one, Heat Shock 70 kDa Protein Activator II, Hsp70 Chemical Co-Chaperone II, YM-08, YM08, YM8

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€219.00

€219.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€219.00

About This Item

Empirical Formula (Hill Notation):
C19H17N3OS2
CAS Number:
Molecular Weight:
367.49
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€219.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

0.6 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

CCN1C(=CC2=CC=CC=N2)SC(=C3N(C4=CC=CC=C4S3)C)C1=O

InChI

1S/C19H17N3OS2/c1-3-22-16(12-13-8-6-7-11-20-13)25-17(18(22)23)19-21(2)14-9-4-5-10-15(14)24-19/h4-12H,3H2,1-2H3

InChI key

UYFLAORTCSDPFM-UHFFFAOYSA-N

General description

A cell-permeable, neutrally charged, pyridinium-to-pyridine substituted YM-01 (Cat. No. 500615) and MKT-077 structural analog that exhibits higher HSP70-binding affinity than its cationic pyridinium counterparts (IC50 = 0.61 µM/YM-08, 3.2 µM/YM-01, and 6.4 µM/MKT-077, respectively, in competitive binding assays using human Hsc70/HSPA8), but reduced tau degradation-promoting potency in cultures (% degradation of p-tau/total tau/24 h = 42/64, 88/89, and 81/80 in HeLaC3 with 30 µM YM-08, MKT-077, or YM-01, respectively) and weaker anti-cancer activity (IC50 against MDA-MB-231/MCF10A/MCF7 in µM = 1.4/3.0/2,2/MKT-077, 2.0/3.3/5.2/YM-01, and 8.5/7.8/10.5/YM-08 with 72 h compound treatment). However, only YM-08, but not YM-01 or MKT-077, exhibits detectable blood-brain-barrier permeability in mice (Cmax = 4 µg/g; t1/2 = 6.8 h; AUCinf = 260 ng·h/g; 6.6 mg/kg via i.v.; 100 µL/mouse in 30% H2O/5% Cremophor/5% EtOH/60% PBS) with much reduced accumulation in kidney in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HSP70
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Miyata, Y., et al. 2013. ACS Chem. Neurosci.4, 930.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service