Skip to Content
Merck
All Photos(1)

Key Documents

KEY444790339

Key Organics / BIONET

tert-butyl N-methyl-N-[(3R)-piperidin-3-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
214.31
MDL number:

Iupac Name

tert-butyl methyl[(3R)-3-piperidinyl]carbamate

Inchi Code

1S/C11H22N2O2/c1-11(2,3)15-10(14)13(4)9-6-5-7-12-8-9/h9,12H,5-8H2,1-4H3/t9-/m1/s1

InChI key

RTXNDTNDOHQMTI-SECBINFHSA-N

storage temp.

Store at 4C

Purity

95%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/LS-1028

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/LS-1028


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service