Skip to Content
Merck
All Photos(1)

Key Documents

KEY370561060

Key Organics / BIONET

(6-phenylpyridin-3-yl)boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
199.02
MDL number:

Iupac Name

6-phenyl-3-pyridinylboronic acid

Inchi Code

1S/C11H10BNO2/c14-12(15)10-6-7-11(13-8-10)9-4-2-1-3-5-9/h1-8,14-15H

InChI key

XSQUPVXOENTCJV-UHFFFAOYSA-N

storage temp.

Store under inert gas

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/BS-2016

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/BS-2016


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service